Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hexb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010422.2 | NP_034552.1 | 2 - 3 | 483 | 80 | |
GenBank mRNA | AK151149.1 | - | 2 - 3 | 420 | 80 | |
AK159134.1 | - | 2 - 3 | 436 | 80 | ||
AK159178.1 | - | 2 - 3 | 457 | 80 | ||
AK160560.1 | - | 2 - 3 | 452 | 80 | ||
AK168825.1 | - | 2 - 3 | 457 | 80 | ||
U07633.1 | - | 2 - 3 | 455 | 80 | ||
Y00964.1 | - | 2 - 3 | 396 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons |