Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mccc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030026.2 | NP_084302.1 | 9 - 10 | 1029 | 103 | |
GenBank mRNA | AK019760.1 | - | 3 - 4 | 210 | 103 | |
AK132265.1 | - | 9 - 10 | 1009 | 103 | ||
AK143233.1 | - | 9 - 10 | 1030 | 103 | ||
AK145564.1 | - | 9 - 10 | 981 | 103 | ||
AK146844.1 | - | 9 - 10 | 989 | 103 | ||
AK146865.1 | - | 9 - 10 | 979 | 103 | ||
BC141193.1 | - | 9 - 10 | 995 | 9055805 | 103 | |
BC141194.1 | - | 9 - 10 | 995 | 9055806 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |