Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178912.3 | NP_849243.2 | 12 - 13 | 2203 | 87 | |
GenBank mRNA | AK050306.1 | - | 12 - 13 | 2204 | 87 | |
AK084697.1 | - | 12 - 13 | 2207 | 87 | ||
AK173211.1 | - | 12 - 13 | 2223 | 87 | ||
BC150785.1 | - | 12 - 13 | 2197 | 9087696 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |