Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178912.3 | NP_849243.2 | 14 - 15 | 4219 | 85 | |
GenBank mRNA | AK050306.1 | - | 14 - 15 | 4220 | 85 | |
AK084697.1 | - | 14 - 15 | 4223 | 85 | ||
AK173211.1 | - | 14 - 15 | 4239 | 85 | ||
BC150785.1 | - | 14 - 15 | 4213 | 9087696 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |