Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbl1xr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030732.3 | NP_109657.2 | 15 - 16 | 1928 | 71 | |
GenBank mRNA | AF268195.1 | - | 15 - 16 | 1709 | 71 | |
AK029595.1 | - | 15 - 16 | 1727 | 71 | ||
AK033347.1 | - | 15 - 16 | 1798 | 71 | ||
AK036064.1 | - | 16 - 17 | 2039 | 71 | ||
AK136255.1 | - | 15 - 16 | 1750 | 71 | ||
BC018512.1 | - | 5 - 6 | 574 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |