Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pappa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085376.2 | NP_001078845.1 | 11 - 12 | 3799 | 61 | |
GenBank mRNA | AK084006.1 | - | 11 - 12 | 3316 | 61 | |
BC094560.1 | - | 1 - 2 | 60 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Mus musculus Probe spans exons |