Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actl6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019673.2 | NP_062647.2 | 10 - 11 | 1260 | 76 | |
GenBank mRNA | AK090154.1 | - | 10 - 11 | 1079 | 76 | |
AK167905.1 | - | 10 - 11 | 1074 | 76 | ||
AK169278.1 | - | 10 - 11 | 1079 | 76 | ||
BC001994.1 | - | 10 - 11 | 1048 | 3491205 | 76 | |
BC094498.1 | - | 10 - 11 | 1076 | 4209491 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |