Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prpf39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK154170.1 | - | 3 - 4 | 1130 | 121 | |
BC014689.1 | - | 3 - 4 | 585 | 121 | ||
BC030904.1 | - | 2 - 3 | 425 | 121 | ||
BC034078.1 | - | 5 - 6 | 817 | 121 | ||
BC076622.1 | - | 2 - 3 | 192 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |