Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Canx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110499.1 | NP_001103969.1 | 8 - 9 | 1092 | 83 | |
NM_001110500.1 | NP_001103970.1 | 8 - 9 | 1060 | 83 | ||
NM_007597.3 | NP_031623.1 | 8 - 9 | 1096 | 83 | ||
GenBank mRNA | AK028279.1 | - | 8 - 9 | 1024 | 83 | |
AK084175.1 | - | 8 - 9 | 1036 | 83 | ||
AK159298.1 | - | 8 - 9 | 1080 | 83 | ||
AK165508.1 | - | 8 - 9 | 995 | 83 | ||
AK166649.1 | - | 8 - 9 | 1073 | 83 | ||
BC012408.1 | - | 8 - 9 | 1014 | 3497769 | 83 | |
BC040244.1 | - | 8 - 9 | 1023 | 2582119 | 83 | |
L18888.1 | - | 7 - 8 | 908 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |