Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sell | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164059.1 | NP_001157531.1 | 5 - 6 | 958 | 87 | |
NM_011346.2 | NP_035476.1 | 6 - 7 | 1066 | 87 | ||
GenBank mRNA | AB353767.1 | - | 5 - 6 | 953 | 87 | |
AB353768.1 | - | 5 - 6 | 953 | 87 | ||
AK137509.1 | - | 6 - 7 | 978 | 87 | ||
AK161754.1 | - | 6 - 7 | 1058 | 87 | ||
AK163066.1 | - | 4 - 5 | 523 | 87 | ||
AK170753.1 | - | 5 - 6 | 959 | 87 | ||
BC052681.1 | - | 6 - 7 | 1050 | 30072021 | 87 | |
M25324.1 | - | 6 - 7 | 1058 | 87 | ||
M36005.1 | - | 6 - 7 | 1004 | 87 | ||
X14772.1 | - | 6 - 7 | 1007 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |