Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdrd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134741.1 | NP_001128213.1 | 4 - 5 | 866 | 76 | |
NM_001277730.1 | NP_001264659.1 | 4 - 5 | 572 | 76 | ||
GenBank mRNA | AK132674.1 | - | 4 - 5 | 852 | 76 | |
AK133026.1 | - | 2 - 3 | 236 | 76 | ||
BC099972.1 | - | 4 - 5 | 572 | 30080156 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |