Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200001566.mm.1 |
NM_145890.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001566.mm.1 |
NM_145890.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001566.mm.1 |
TC1200000200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200001570.mm.1 |
NM_001161406.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001570.mm.1 |
NM_001161406.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001570.mm.1 |
NM_145890.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001570.mm.1 |
NM_145890.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001570.mm.1 |
TC1200000200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200001571.mm.1 |
NM_001161406.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001571.mm.1 |
NM_001161406.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001571.mm.1 |
NM_145890.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001571.mm.1 |
NM_145890.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001571.mm.1 |
TC1200000200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200001567.mm.1 |
NM_145890.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001567.mm.1 |
NM_145890.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001567.mm.1 |
TC1200000200.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200001569.mm.1 |
NM_001161406.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001569.mm.1 |
NM_001161406.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001569.mm.1 |
NM_145890.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001569.mm.1 |
NM_145890.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200001569.mm.1 |
TC1200000200.mm.1 |
MTA TC |