Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081307.1 | NP_001074776.1 | 20 - 21 | 2787 | 92 | |
GenBank mRNA | AK039445.1 | - | 8 - 9 | 848 | 92 | |
BC145746.1 | - | 20 - 21 | 2698 | 40131049 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |