Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srgap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081011.2 | NP_001074480.2 | 6 - 7 | 1420 | 76 | |
GenBank mRNA | BC030457.1 | - | 5 - 6 | 727 | 76 | |
BC151081.1 | - | 5 - 6 | 1038 | 9087994 | 76 | |
BC151082.1 | - | 5 - 6 | 1038 | 9087995 | 76 | |
BC158055.1 | - | 5 - 6 | 1036 | 9088163 | 76 | |
BC172152.1 | - | 5 - 6 | 1038 | 9054822 | 76 | |
BE653295.1 | - | 2 - 3 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |