Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281965.1 | NP_001268894.1 | 7 - 8 | 837 | 88 | |
NM_001281966.1 | NP_001268895.1 | 7 - 8 | 828 | 88 | ||
NM_001281967.1 | NP_001268896.1 | 7 - 8 | 819 | 88 | ||
NM_001281968.1 | NP_001268897.1 | 7 - 8 | 816 | 88 | ||
NM_010583.3 | NP_034713.2 | 7 - 8 | 819 | 88 | ||
GenBank mRNA | AK153613.1 | - | 7 - 8 | 775 | 88 | |
BC028478.1 | - | 7 - 8 | 761 | 88 | ||
BC128374.1 | - | 7 - 8 | 745 | 40111687 | 88 | |
BC128375.1 | - | 7 - 8 | 742 | 40111688 | 88 | |
D14042.1 | - | 7 - 8 | 759 | 88 | ||
L00619.1 | - | 7 - 8 | 824 | 88 | ||
L05631.1 | - | 7 - 8 | 785 | 88 | ||
L10628.1 | - | 7 - 8 | 787 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Protein Kinase 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |