Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136070.1 | NP_001129542.1 | 14 - 15 | 1803 | 106 | |
GenBank mRNA | AK030312.1 | - | 14 - 15 | 1814 | 106 | |
AK046539.1 | - | 14 - 15 | 1815 | 106 | ||
AK083638.1 | - | 14 - 15 | 1763 | 106 | ||
AK139052.1 | - | 14 - 15 | 1803 | 106 | ||
AK161269.1 | - | 14 - 15 | 1786 | 106 | ||
AK166085.1 | - | 14 - 15 | 1816 | 106 | ||
AK167980.1 | - | 14 - 15 | 1799 | 106 | ||
AK168120.1 | - | 14 - 15 | 1769 | 106 | ||
AK170252.1 | - | 14 - 15 | 1830 | 106 | ||
AK170448.1 | - | 14 - 15 | 1815 | 106 | ||
AK171115.1 | - | 14 - 15 | 1781 | 106 | ||
BC023709.1 | - | 14 - 15 | 1756 | 106 | ||
BC024501.1 | - | 9 - 10 | 1117 | 106 | ||
BC025036.1 | - | 14 - 15 | 1759 | 4935223 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |