Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgef1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029004.1 | NP_083280.1 | 12 - 13 | 1443 | 79 | |
GenBank mRNA | AK018593.1 | - | 12 - 13 | 1432 | 79 | |
AK039330.1 | - | 1807 | 79 | |||
AK047088.1 | - | 11 - 12 | 1322 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |