Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290759.1 | NP_001277688.1 | 1 - 2 | 197 | 99 | |
NM_001290760.1 | NP_001277689.1 | 1 - 2 | 197 | 99 | ||
NM_029745.2 | NP_084021.2 | 1 - 2 | 197 | 99 | ||
GenBank mRNA | AK035457.1 | - | 1 - 2 | 162 | 99 | |
AK049999.1 | - | 1 - 2 | 157 | 99 | ||
AK147964.1 | - | 1 - 2 | 162 | 99 | ||
AK166472.1 | - | 1 - 2 | 180 | 100 | ||
AK172999.1 | - | 1 - 2 | 137 | 99 | ||
BC062928.1 | - | 1 - 2 | 184 | 6854669 | 99 | |
BC065080.1 | - | 1 - 2 | 176 | 6826791 | 99 | |
BC065130.1 | - | 1 - 2 | 142 | 99 | ||
BY207144.1 | - | 1 - 2 | 197 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |