Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF005423.1 | - | 2 - 3 | 85 | ||
AK013974.2 | - | 3 - 4 | 380 | 85 | ||
AK161739.1 | - | 1 - 2 | 1634 | 85 | ||
AY653308.1 | - | 4 - 5 | 515 | 85 | ||
BC002220.1 | - | 1 - 2 | 181 | 3490616 | 85 | |
BC012675.1 | - | 5 - 6 | 647 | 3982920 | 85 | |
CT010293.1 | - | 4 - 5 | 547 | 85 | ||
U94846.1 | - | 5 - 6 | 644 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons |