Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011891.4 | NP_036021.1 | 2 - 3 | 239 | 86 | |
GenBank mRNA | AB024923.1 | - | 2 - 3 | 460 | 86 | |
AK040974.1 | - | 2 - 3 | 241 | 86 | ||
BC130019.1 | - | 2 - 3 | 229 | 40140624 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |