Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slu7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148673.3 | NP_683514.2 | 5 - 6 | 594 | 77 | |
NM_198936.1 | NP_945174.1 | 4 - 5 | 515 | 77 | ||
GenBank mRNA | AK029117.1 | - | 4 - 5 | 485 | 77 | |
AK046262.1 | - | 4 - 5 | 486 | 77 | ||
AK047597.1 | - | 5 - 6 | 582 | 77 | ||
AK049178.1 | - | 4 - 5 | 484 | 77 | ||
BC013810.1 | - | 2 - 3 | 114 | 77 | ||
BC025870.1 | - | 5 - 6 | 573 | 77 | ||
BC060954.1 | - | 4 - 5 | 483 | 77 | ||
BC082780.1 | - | 5 - 6 | 574 | 77 | ||
BC106099.1 | - | 4 - 5 | 496 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |