Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300001447.mm.1 |
NM_025978.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001447.mm.1 |
NM_027619.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001447.mm.1 |
TC0300000233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001459.mm.1 |
NM_025978.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001459.mm.1 |
NM_027619.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001459.mm.1 |
TC0300000233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001453.mm.1 |
NM_025978.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001453.mm.1 |
NM_027619.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001453.mm.1 |
TC0300000233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001454.mm.1 |
NM_025978.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001454.mm.1 |
NM_027619.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001454.mm.1 |
TC0300000233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001446.mm.1 |
NM_025978.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001446.mm.1 |
NM_027619.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001446.mm.1 |
TC0300000233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300001458.mm.1 |
NM_025978.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001458.mm.1 |
NM_027619.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300001458.mm.1 |
TC0300000233.mm.1 |
MTA TC |