Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007628.3 | NP_031654.2 | 4 - 5 | 641 | 68 | |
GenBank mRNA | AK077114.1 | - | 4 - 5 | 647 | 68 | |
BC120518.1 | - | 3 - 4 | 556 | 40129441 | 68 | |
BC125436.1 | - | 3 - 4 | 556 | 40129951 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |