Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201357.2 | NP_958745.2 | 6 - 7 | 790 | 64 | |
GenBank mRNA | AK150308.1 | - | 6 - 7 | 791 | 64 | |
AK151830.1 | - | 6 - 7 | 791 | 64 | ||
BC031458.1 | - | 6 - 7 | 753 | 4191954 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |