Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027534.2 | NP_081810.1 | 6 - 7 | 702 | 67 | |
GenBank mRNA | AK018155.1 | - | 7 - 8 | 677 | 67 | |
AK170064.1 | - | 6 - 7 | 585 | 67 | ||
AY634684.1 | - | 6 - 7 | 611 | 67 | ||
BC023820.1 | - | 6 - 7 | 692 | 5354804 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |