Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sctr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012322.2 | NP_001012322.2 | 12 - 13 | 1459 | 74 | |
GenBank mRNA | AK032652.1 | - | 12 - 13 | 1459 | 74 | |
BC089355.1 | - | 11 - 12 | 1360 | 30551620 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Mammalian Gene Collection Mus musculus Probe spans exons |