Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nckap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081756.1 | NP_001075225.1 | 15 - 16 | 5688 | 77 | |
NM_172484.3 | NP_766072.2 | 13 - 14 | 5549 | 77 | ||
GenBank mRNA | AY957601.1 | - | 10 - 11 | 4916 | 77 | |
AY957602.1 | - | 11 - 12 | 5108 | 77 | ||
BC066095.1 | - | 13 - 14 | 5549 | 6856184 | 77 | |
BC079551.1 | - | 13 - 14 | 5549 | 6856184 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |