Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco4c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172658.3 | NP_766246.1 | 5 - 6 | 1066 | 91 | |
GenBank mRNA | AK049253.1 | - | 5 - 6 | 1066 | 91 | |
AK049944.1 | - | 1031 | 91 | |||
AY654588.1 | - | 5 - 6 | 1066 | 91 | ||
BC119022.1 | - | 5 - 6 | 1025 | 8733771 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |