Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK013209.1 | - | 5 - 6 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |