Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168321.1 | NP_001161793.1 | 7 - 8 | 3808 | 84 | |
NM_029510.3 | NP_083786.2 | 8 - 9 | 4633 | 84 | ||
NM_175044.3 | NP_778209.2 | 7 - 8 | 4579 | 84 | ||
NM_175045.3 | NP_778210.2 | 8 - 9 | 4531 | 84 | ||
NM_175046.3 | NP_778211.2 | 7 - 8 | 4477 | 84 | ||
GenBank mRNA | AK129398.1 | - | 7 - 8 | 3808 | 84 | |
AY161170.1 | - | 8 - 9 | 3948 | 84 | ||
AY161171.1 | - | 7 - 8 | 3894 | 84 | ||
AY161172.1 | - | 8 - 9 | 3846 | 84 | ||
AY161173.1 | - | 7 - 8 | 3792 | 84 | ||
BC058656.1 | - | 8 - 9 | 4032 | 6412868 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |