Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039515.1 | NP_001034604.1 | 1 - 2 | 151 | 108 | |
GenBank mRNA | AK006286.1 | - | 1 - 2 | 91 | 107 | |
AK161281.1 | - | 1 - 2 | 178 | 108 | ||
CJ164717.1 | - | 1 - 2 | 157 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons |