Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bzw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025840.3 | NP_080116.2 | 10 - 11 | 1279 | 85 | |
GenBank mRNA | AB244988.1 | - | 10 - 11 | 1195 | 85 | |
AK003229.1 | - | 10 - 11 | 1274 | 85 | ||
AK011253.1 | - | 10 - 11 | 1235 | 85 | ||
AK088108.1 | - | 10 - 11 | 1210 | 85 | ||
AK160544.1 | - | 10 - 11 | 1252 | 85 | ||
BC013060.1 | - | 10 - 11 | 1215 | 3482251 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |