Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145507.2 | NP_663482.2 | 1 - 2 | 260 | 117 | |
NM_177445.5 | NP_803228.2 | 1 - 2 | 260 | 117 | ||
GenBank mRNA | AK077533.1 | - | 1 - 2 | 183 | 117 | |
AK077820.1 | - | 1 - 2 | 214 | 117 | ||
AK144470.1 | - | 1 - 2 | 205 | 117 | ||
AK146350.1 | - | 1 - 2 | 204 | 117 | ||
AK151027.1 | - | 1 - 2 | 204 | 117 | ||
AK151897.1 | - | 1 - 2 | 171 | 117 | ||
AK152304.1 | - | 1 - 2 | 171 | 117 | ||
AK152562.1 | - | 1 - 2 | 171 | 117 | ||
AK153256.1 | - | 1 - 2 | 171 | 117 | ||
AK153299.1 | - | 1 - 2 | 171 | 117 | ||
AK166009.1 | - | 1 - 2 | 171 | 117 | ||
AK169247.1 | - | 1 - 2 | 171 | 117 | ||
AK169716.1 | - | 1 - 2 | 148 | 117 | ||
BB576692.2 | - | 1 - 2 | 214 | 117 | ||
BC008638.1 | - | 1 - 2 | 158 | 3586278 | 117 | |
BY276340.1 | - | 1 - 2 | 261 | 117 | ||
BY308833.1 | - | 1 - 2 | 258 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons |