Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019510.2 | NP_062383.2 | 10 - 11 | 2697 | 155 | |
GenBank mRNA | AF190645.1 | - | 9 - 10 | 2287 | 155 | |
AK032317.1 | - | 10 - 11 | 2698 | 155 | ||
AK080619.1 | - | 10 - 11 | 2685 | 155 | ||
BC108988.2 | - | 9 - 10 | 2286 | 155 | ||
BC108989.2 | - | 9 - 10 | 2287 | 40047766 | 155 | |
FJ207475.1 | - | 9 - 10 | 2287 | 155 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Ion Channel Mammalian Gene Collection Mus musculus Probe spans exons |