Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankmy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172850.3 | NP_766438.2 | 5 - 6 | 1040 | 71 | |
GenBank mRNA | AK029700.1 | - | 5 - 6 | 1042 | 71 | |
BC120887.1 | - | 4 - 5 | 876 | 40060916 | 71 | |
BC120888.1 | - | 4 - 5 | 876 | 40060917 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |