Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ezh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AB004817.1 | - | 11 - 12 | 1327 | 112 | |
AF483490.1 | - | 9 - 10 | 1216 | 112 | ||
AF483491.1 | - | 9 - 10 | 1216 | 112 | ||
AK045374.1 | - | 11 - 12 | 1315 | 112 | ||
AK129004.1 | - | 10 - 11 | 1287 | 112 | ||
AK138942.1 | - | 11 - 12 | 1310 | 112 | ||
AK140694.1 | - | 11 - 12 | 1334 | 112 | ||
AK154565.1 | - | 11 - 12 | 1307 | 112 | ||
AK164192.1 | - | 11 - 12 | 1326 | 112 | ||
BC007135.1 | - | 10 - 11 | 1287 | 3600884 | 112 | |
U60453.1 | - | 11 - 12 | 1315 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |