Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100003317.mm.1 |
NM_148673.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003317.mm.1 |
NM_148673.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003317.mm.1 |
TC1100000426.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100003320.mm.1 |
NM_148673.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003320.mm.1 |
NM_148673.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003320.mm.1 |
NM_198936.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003320.mm.1 |
NM_198936.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003320.mm.1 |
TC1100000426.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100003319.mm.1 |
NM_148673.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003319.mm.1 |
NM_148673.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003319.mm.1 |
NM_198936.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003319.mm.1 |
NM_198936.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003319.mm.1 |
TC1100000426.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100003315.mm.1 |
NM_148673.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003315.mm.1 |
NM_148673.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003315.mm.1 |
NM_198936.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003315.mm.1 |
NM_198936.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100003315.mm.1 |
TC1100000426.mm.1 |
MTA TC |