Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033156.4 | NP_001028328.2 | 2 - 3 | 728 | 115 | |
GenBank mRNA | BC020022.1 | - | 2 - 3 | 697 | 115 | |
BC025217.1 | - | 2 - 3 | 308 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |