Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gli2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081125.1 | NP_001074594.1 | 13 - 14 | 2598 | 114 | |
GenBank mRNA | BC082604.1 | - | 13 - 14 | 2539 | 30672479 | 114 |
BC085190.1 | - | 13 - 14 | 2514 | 30694633 | 114 | |
X99104.1 | - | 13 - 14 | 2358 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |