Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mael | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175296.4 | NP_780505.1 | 1 - 2 | 214 | 86 | |
GenBank mRNA | AK077103.1 | - | 1 - 2 | 211 | 86 | |
AK165801.1 | - | 1 - 2 | 211 | 86 | ||
BC050800.1 | - | 1 - 2 | 214 | 6744032 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |