Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Golt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026680.4 | NP_080956.1 | 2 - 3 | 183 | 70 | |
GenBank mRNA | AK002581.1 | - | 2 - 3 | 191 | 70 | |
AK141378.1 | - | 2 - 3 | 184 | 70 | ||
AY707855.1 | - | 2 - 3 | 206 | 70 | ||
BC024448.1 | - | 2 - 3 | 206 | 4974299 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |