Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100004275.mm.1 |
NM_173372.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004275.mm.1 |
NM_173372.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004275.mm.1 |
TC1100000549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004279.mm.1 |
NM_173372.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004279.mm.1 |
NM_173372.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004279.mm.1 |
TC1100000549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004280.mm.1 |
NM_173372.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004280.mm.1 |
NM_173372.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004280.mm.1 |
TC1100000549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004273.mm.1 |
NM_173372.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004273.mm.1 |
NM_173372.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004273.mm.1 |
TC1100000549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004282.mm.1 |
NM_173372.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004282.mm.1 |
NM_173372.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004282.mm.1 |
TC1100000549.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100004278.mm.1 |
NM_173372.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004278.mm.1 |
NM_173372.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100004278.mm.1 |
TC1100000549.mm.1 |
MTA TC |