Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcp4l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025557.1 | NP_079833.1 | 1 - 2 | 259 | 105 | |
GenBank mRNA | AK002772.1 | - | 1 - 2 | 230 | 105 | |
AK032164.1 | - | 1 - 2 | 229 | 105 | ||
AK075738.1 | - | 1 - 2 | 229 | 105 | ||
AY304481.1 | - | 1 - 2 | 230 | 105 | ||
BC116208.1 | - | 1 - 2 | 289 | 106 | ||
BC126965.1 | - | 1 - 2 | 289 | 106 | ||
BY238909.1 | - | 1 - 2 | 259 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |