Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Swt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025819.4 | NP_080095.3 | 14 - 15 | 2182 | 124 | |
GenBank mRNA | AK004803.2 | - | 14 - 15 | 2129 | 124 | |
BC060204.1 | - | 14 - 15 | 2122 | 6855442 | 124 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |