Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172643.5 | NP_766231.2 | 5 - 6 | 1700 | 129 | |
GenBank mRNA | AK034756.1 | - | 4 - 5 | 797 | 129 | |
AK036544.1 | - | 5 - 6 | 1699 | 129 | ||
AK079719.1 | - | 1 - 2 | 58 | 129 | ||
AK163302.1 | - | 4 - 5 | 973 | 129 | ||
BC046465.1 | - | 4 - 5 | 869 | 129 | ||
BC150859.1 | - | 4 - 5 | 1039 | 9087770 | 129 | |
BC172012.1 | - | 5 - 6 | 965 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |