Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Depdc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178683.4 | NP_848798.1 | 4 - 5 | 659 | 69 | |
GenBank mRNA | AK036550.1 | - | 4 - 5 | 618 | 69 | |
AK043283.1 | - | 616 | 69 | |||
AK077676.1 | - | 4 - 5 | 660 | 69 | ||
AK083480.1 | - | 4 - 5 | 655 | 69 | ||
BC152330.1 | - | 4 - 5 | 664 | 40141538 | 69 | |
BC152331.1 | - | 4 - 5 | 664 | 40141540 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |