Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rapgefl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080925.1 | NP_001074394.1 | 3 - 4 | 1099 | 90 | |
GenBank mRNA | BC080279.1 | - | 2 - 3 | 167 | 90 | |
BC130266.1 | - | 4 - 5 | 375 | 40143028 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |