Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009634.6 | NP_033764.2 | 2 - 3 | 420 | 88 | |
GenBank mRNA | AK049372.1 | - | 2 - 3 | 392 | 88 | |
AK051111.1 | - | 2 - 3 | 392 | 88 | ||
AK152092.1 | - | 2 - 3 | 392 | 88 | ||
AK166019.1 | - | 2 - 3 | 388 | 88 | ||
AK168906.1 | - | 2 - 3 | 385 | 88 | ||
AK171725.1 | - | 2 - 3 | 401 | 88 | ||
BC020187.1 | - | 2 - 3 | 396 | 4913637 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |