Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025730.3 | NP_080006.3 | 35 - 36 | 5229 | 106 | |
GenBank mRNA | AY792512.1 | - | 35 - 36 | 5172 | 106 | |
BC034074.1 | - | 1 - 2 | 112 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |