Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193309.1 | NP_001180238.1 | 5 - 6 | 959 | 104 | |
NM_029413.4 | NP_083689.2 | 5 - 6 | 959 | 104 | ||
GenBank mRNA | AK017472.1 | - | 5 - 6 | 928 | 104 | |
BC098483.1 | - | 5 - 6 | 914 | 30022418 | 104 | |
BC132153.1 | - | 5 - 6 | 813 | 40130430 | 104 | |
BC132497.1 | - | 5 - 6 | 813 | 40130774 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |